Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGL
Entrez Gene ID
507080
Gene Name
phosphatidylinositol glycan anchor biosynthesis class L
Gene Aliases
-
Chromosome Location
Chr.19: 33276803 - 33326729 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100311.2 NP_001093781.1 2 - 3 351 139
GenBank mRNA BC149604.1 - 2 - 3 353 139

View Full Product Details