Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINE3
Entrez Gene ID
513955
Gene Name
serpin family E member 3
Gene Aliases
-
Chromosome Location
Chr.12: 20784861 - 20816359 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100359.3 NP_001093829.1 5 - 6 1059 90

View Full Product Details