Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A1
Entrez Gene ID
519461
Gene Name
solute carrier family 22 member 1
Gene Aliases
OCT1
Chromosome Location
Chr.9: 96309292 - 96352834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101098.2 NP_001094568.1 4 - 5 865 91
GenBank mRNA BC151505.1 - 4 - 5 866 91

View Full Product Details