Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT20
Entrez Gene ID
505214
Gene Name
keratin 20
Gene Aliases
-
Chromosome Location
Chr.19: 41065645 - 41075614 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101851.2 NP_001095321.1 1 - 2 443 71
GenBank mRNA BC149955.1 - 1 - 2 445 71

View Full Product Details