The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101937.1 | NP_001095407.1 | 5 - 6 | 641 | 124 |
GenBank mRNA | BC149229.1 | - | 5 - 6 | 641 | 124 |