Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEPSIN
Entrez Gene ID
512349
Gene Name
TEPSIN adaptor related protein complex 4 accessory protein
Gene Aliases
C19H17orf56, ENTHD2
Chromosome Location
Chr.19: 51483093 - 51494286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101954.1 NP_001095424.1 2 - 3 398 145
GenBank mRNA BC149149.1 - 2 - 3 398 145

View Full Product Details