Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STYXL1
Entrez Gene ID
513107
Gene Name
serine/threonine/tyrosine interacting like 1
Gene Aliases
-
Chromosome Location
Chr.25: 34222291 - 34255029 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101968.2 NP_001095438.1 2 - 3 128 123
GenBank mRNA BC151531.1 - 2 - 3 129 123

View Full Product Details