Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HERC5
Entrez Gene ID
514373
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
-
Chromosome Location
Chr.6: 36257276 - 36302337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101995.1 NP_001095465.1 4 - 5 778 122
GenBank mRNA BC149081.1 - 4 - 5 778 122

View Full Product Details