Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HERC5
Entrez Gene ID
514373
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
-
Chromosome Location
Chr.6: 36257276 - 36302337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101995.1 NP_001095465.1 5 - 6 875 107
GenBank mRNA BC149081.1 - 5 - 6 875 107

View Full Product Details