Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URB2
Entrez Gene ID
514623
Gene Name
URB2 ribosome biogenesis homolog
Gene Aliases
-
Chromosome Location
Chr.28: 1577579 - 1604956 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102004.1 NP_001095474.1 7 - 8 4302 66
GenBank mRNA BC148096.1 - 7 - 8 4302 66

View Full Product Details