Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPAG8
Entrez Gene ID
514627
Gene Name
sperm associated antigen 8
Gene Aliases
-
Chromosome Location
Chr.8: 59974964 - 59977598 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102005.2 NP_001095475.1 4 - 5 1533 78
GenBank mRNA BC149514.1 - 4 - 5 1534 78

View Full Product Details