Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KANK4
Entrez Gene ID
519027
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
-
Chromosome Location
Chr.3: 83258461 - 83334028 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC149084.1 - 3 - 4 470 113

View Full Product Details