Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP69
Entrez Gene ID
521549
Gene Name
ZFP69 zinc finger protein
Gene Aliases
ZNF642
Chromosome Location
Chr.3: 105656766 - 105675329 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102063.1 NP_001095533.1 4 - 5 690 147
GenBank mRNA BC151573.1 - 4 - 5 690 147

View Full Product Details