The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102088.2 | NP_001095558.1 | 5 - 6 | 652 | 103 |
GenBank mRNA | BC151501.1 | - | 5 - 6 | 653 | 103 |