Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDAD1
Entrez Gene ID
507675
Gene Name
SDA1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 90809711 - 90841174 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102489.1 NP_001095959.1 17 - 18 1570 140
GenBank mRNA BC140502.1 - 17 - 18 1570 140

View Full Product Details