Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO17
Entrez Gene ID
510874
Gene Name
F-box protein 17
Gene Aliases
-
Chromosome Location
Chr.18: 48642188 - 48674144 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103239.2 NP_001096709.2 1 - 2 126 125
GenBank mRNA BC149102.1 - 1 - 2 126 125

View Full Product Details