Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAT9
Entrez Gene ID
514377
Gene Name
N-acetyltransferase 9 (putative)
Gene Aliases
-
Chromosome Location
Chr.19: 56647397 - 56654164 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103246.1 NP_001096716.1 4 - 5 729 86
GenBank mRNA BC149879.1 - 4 - 5 729 86

View Full Product Details