Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACER1
Entrez Gene ID
518021
Gene Name
alkaline ceramidase 1
Gene Aliases
ASAH3
Chromosome Location
Chr.7: 18076214 - 18129207 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103254.1 NP_001096724.1 2 - 3 216 72
GenBank mRNA BC149050.1 - 2 - 3 216 72

View Full Product Details