Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX12
Entrez Gene ID
520739
Gene Name
sorting nexin 12
Gene Aliases
-
Chromosome Location
Chr.X: 79555820 - 79618906 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103257.1 NP_001096727.1 2 - 3 284 75
GenBank mRNA BC150127.1 - 2 - 3 284 75

View Full Product Details