Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D10A
Entrez Gene ID
511933
Gene Name
TBC1 domain family member 10A
Gene Aliases
-
Chromosome Location
Chr.17: 69307674 - 69336442 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104974.1 NP_001098444.1 2 - 3 377 72
GenBank mRNA BC151382.1 - 2 - 3 377 72

View Full Product Details