Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF6
Entrez Gene ID
523017
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 6
Gene Aliases
C14H8orf38
Chromosome Location
Chr.14: 69265272 - 69301597 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105389.1 NP_001098859.1 8 - 9 877 130
GenBank mRNA BC123431.1 - 8 - 9 877 130

View Full Product Details