Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL2
Entrez Gene ID
512019
Gene Name
angiopoietin like 2
Gene Aliases
-
Chromosome Location
Chr.11: 97835686 - 97872064 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109814.1 NP_001103284.1 2 - 3 1036 67
GenBank mRNA EU599225.1 - 2 - 3 1026 67

View Full Product Details