Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAT3
Entrez Gene ID
516262
Gene Name
G protein subunit alpha transducin 3
Gene Aliases
-
Chromosome Location
Chr.4: 40574285 - 40629320 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109982.1 NP_001103452.1 4 - 5 542 95

View Full Product Details