Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBIS
Entrez Gene ID
510007
Gene Name
ribosomal biogenesis factor
Gene Aliases
C14H8orf59
Chromosome Location
Chr.14: 77391690 - 77396788 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110068.2 NP_001103538.1 2 - 3 178 148
GenBank mRNA BC108159.1 - 1 - 1 179 148

View Full Product Details