Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG4
Entrez Gene ID
516353
Gene Name
BAG cochaperone 4
Gene Aliases
-
Chromosome Location
Chr.27: 33349063 - 33374595 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110532.2 NP_001104002.1 1 - 2 323 123
GenBank mRNA BT025395.1 - 1 - 2 267 123
BT030482.1 - 1 - 2 267 123

View Full Product Details