Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF3B
Entrez Gene ID
508404
Gene Name
kinesin family member 3B
Gene Aliases
-
Chromosome Location
Chr.13: 61747749 - 61787774 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110788.1 NP_001104258.1 1 - 2 89 75

View Full Product Details