Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTL
Entrez Gene ID
514559
Gene Name
tubulin tyrosine ligase
Gene Aliases
-
Chromosome Location
Chr.11: 46209289 - 46242645 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113233.1 NP_001106704.1 6 - 7 1020 70

View Full Product Details