The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113718.2 | NP_001107190.1 | 2 - 3 | 294 | 138 |
GenBank mRNA | AY911344.1 | - | 2 - 3 | 288 | 138 |
BC109531.1 | - | 2 - 3 | 295 | 138 |