Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDGFB
Entrez Gene ID
540106
Gene Name
platelet derived growth factor subunit B
Gene Aliases
-
Chromosome Location
Chr.5: 110665513 - 110686525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017953.2 NP_001017953.2 6 - 7 750 156
GenBank mRNA BC151383.1 - 6 - 7 750 156
BT021749.1 - 6 - 7 790 156

View Full Product Details