Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP10
Entrez Gene ID
541175
Gene Name
dual specificity phosphatase 10
Gene Aliases
-
Chromosome Location
Chr.16: 25186200 - 25227307 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034725.2 NP_001029897.2 1 - 2 81 74
GenBank mRNA BC122694.1 - 1 - 2 81 74
BT021576.1 - 1 - 2 89 74

View Full Product Details