Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WTAP
- Entrez Gene ID
-
532996
- Gene Name
- WT1 associated protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 95996793 - 96023437 on Build ARS-UCD1.2
- Gene Symbol
- LOC104975673
- Entrez Gene ID
-
104975673
- Gene Name
- pre-mRNA-splicing regulator WTAP
- Gene Aliases
-
-
- Chromosome Location
-
Chr.23: 30493361 - 30514886 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC149777.1 |
- |
2 - 2 |
430 |
92 |
View Full Product Details