Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL2BP
Entrez Gene ID
613462
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Gene Aliases
-
Chromosome Location
Chr.18: 25251866 - 25257893 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076274.2 NP_001069742.2 5 - 6 667 96
GenBank mRNA BC108166.1 - 5 - 6 483 96
BC134728.1 - 5 - 6 667 96

View Full Product Details