Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YBX3
Entrez Gene ID
533297
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA
Chromosome Location
Chr.5: 98853392 - 98879071 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098074.1 NP_001091543.1 9 - 10 1185 107
GenBank mRNA BC140644.1 - 9 - 10 1185 107

View Full Product Details