Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFM3
Entrez Gene ID
538499
Gene Name
olfactomedin 3
Gene Aliases
-
Chromosome Location
Chr.3: 41271715 - 41495292 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098094.2 NP_001091563.1 2 - 3 643 140
GenBank mRNA BC140649.1 - 2 - 3 644 140

View Full Product Details