Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTF3L4
Entrez Gene ID
539753
Gene Name
basic transcription factor 3 like 4
Gene Aliases
-
Chromosome Location
Chr.3: 94255745 - 94277771 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098106.2 NP_001091575.1 4 - 5 458 97
GenBank mRNA BC112459.1 - 4 - 5 459 97

View Full Product Details