Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBED8
Entrez Gene ID
539846
Gene Name
zinc finger BED-type containing 8
Gene Aliases
C7H5orf54
Chromosome Location
Chr.7: 71977547 - 71983299 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098107.2 NP_001091576.1 1 - 2 365 127
GenBank mRNA BC140632.1 - 1 - 2 366 127

View Full Product Details