Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYPA
Entrez Gene ID
617836
Gene Name
glycophorin A (MNS blood group)
Gene Aliases
gpa
Chromosome Location
Chr.17: 14040368 - 14066548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098141.1 NP_001091610.1 5 - 6 321 96
GenBank mRNA AB300617.1 - 5 - 6 321 96

View Full Product Details