Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX24
Entrez Gene ID
529613
Gene Name
DEAD-box helicase 24
Gene Aliases
-
Chromosome Location
Chr.21: 58661570 - 58680062 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099045.1 NP_001092515.1 2 - 3 782 71
GenBank mRNA BC146184.1 - 2 - 3 782 71

View Full Product Details