Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOR1A
Entrez Gene ID
533699
Gene Name
torsin family 1 member A
Gene Aliases
-
Chromosome Location
Chr.11: 100196653 - 100205865 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099058.2 NP_001092528.1 3 - 4 682 111
GenBank mRNA BC142387.1 - 3 - 4 683 111

View Full Product Details