Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC29
Entrez Gene ID
536009
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.17: 11330412 - 11607518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099073.2 NP_001092543.1 2 - 3 108 82
GenBank mRNA BC142073.1 - 2 - 3 110 82

View Full Product Details