Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POGK
Entrez Gene ID
539790
Gene Name
pogo transposable element derived with KRAB domain
Gene Aliases
-
Chromosome Location
Chr.3: 2051950 - 2069656 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099100.1 NP_001092570.1 2 - 3 244 112
GenBank mRNA BC142233.1 - 2 - 3 244 112

View Full Product Details