Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXD10
Entrez Gene ID
540251
Gene Name
homeobox D10
Gene Aliases
-
Chromosome Location
Chr.2: 20793703 - 20796809 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099105.1 NP_001092575.1 1 - 2 783 71
GenBank mRNA BC142145.1 - 1 - 2 783 71

View Full Product Details