Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2W
Entrez Gene ID
613423
Gene Name
ubiquitin conjugating enzyme E2 W
Gene Aliases
-
Chromosome Location
Chr.14: 37075725 - 37166741 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099122.3 NP_001092592.1 5 - 6 482 136
GenBank mRNA BC146162.1 - 5 - 6 451 136
FE011537.1 - 5 - 6 482 136

View Full Product Details