Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM70
Entrez Gene ID
613774
Gene Name
transmembrane protein 70
Gene Aliases
-
Chromosome Location
Chr.14: 37225965 - 37233935 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099126.2 NP_001092596.1 2 - 3 402 112
GenBank mRNA BC146138.1 - 2 - 3 403 112

View Full Product Details