Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM41B
Entrez Gene ID
616372
Gene Name
transmembrane protein 41B
Gene Aliases
-
Chromosome Location
Chr.15: 43311264 - 43339038 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099156.1 NP_001092626.1 6 - 7 877 92
GenBank mRNA BC142213.1 - 6 - 7 877 92

View Full Product Details