Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENO2
Entrez Gene ID
526006
Gene Name
enolase 2
Gene Aliases
-
Chromosome Location
Chr.5: 103538376 - 103546537 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101125.1 NP_001094595.1 1 - 2 118 90
GenBank mRNA BC150078.1 - 1 - 2 118 90

View Full Product Details