Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC42
Entrez Gene ID
536427
Gene Name
coiled-coil domain containing 42
Gene Aliases
-
Chromosome Location
Chr.19: 28254549 - 28267354 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101166.2 NP_001094636.1 5 - 6 923 81
GenBank mRNA BC149908.1 - 5 - 6 924 81

View Full Product Details