Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF8
Entrez Gene ID
539938
Gene Name
TATA-box binding protein associated factor 8
Gene Aliases
-
Chromosome Location
Chr.23: 15794632 - 15814406 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101200.2 NP_001094670.1 5 - 6 504 88
GenBank mRNA BC151255.1 - 5 - 6 505 88

View Full Product Details