Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFAP4
Entrez Gene ID
540757
Gene Name
transcription factor AP-4
Gene Aliases
-
Chromosome Location
Chr.25: 3443802 - 3456033 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101215.1 NP_001094685.1 6 - 7 1086 64
GenBank mRNA BC148098.1 - 6 - 7 1086 64

View Full Product Details