Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNLT3
Entrez Gene ID
613349
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
-
Chromosome Location
Chr.X: 105545530 - 105556552 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101220.1 NP_001094690.1 3 - 4 213 93
GenBank mRNA BC148062.1 - 3 - 4 213 93

View Full Product Details